missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.9%, for biochemistry, AcroSeal™

CAS: 67-56-1 | CH4O | 32.04 g/mol

$145.39 - $174.20

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124790010
SDS View Documents
Thermo Scientific Chemicals
124790010
1 L AcroSeal™ Glass Bottle
Each for $145.39
Only null left
 
AC124790025
SDS View Documents
Thermo Scientific Chemicals
124790025
2.5 L AcroSeal™ Glass Bottle
Each for $174.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

-98.0°C
0.01 max. at 260nm, 0.01 max. at 400nm, 0.05 max. at 240nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 0.6 max. at 210nm, 1 max. at 207nm
64.7°C
12°C
1.3280 to 1.3300
0.791
CH3OH
0.55 mPa.s (20°C)
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
32.04
CHEBI:17790
99.9%
Methanol
67-56-1
0.7910g/mL
1 L
AcroSeal™ Glass Bottle
99.9%
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Biochemical
Videos
Safety and Handling

Safety and Handling

Danger

SDS
Documents

Documents

RUO – Research Use Only