Learn More
Research Products Corporation Methanol [Reagent Grade], 4 Liters, 4 Bottles per Case
Catalog No. 50999162
Encompass
Colorless liquid, Flammable, Flash pt. 12°C, b.p. 64-65°C
WARNING! This product can expose you to Methanol, a chemical which is known to the State of California to cause Developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50999162 Supplier Research Products Corporation Supplier No. RPI 400031CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Colorless liquid, Flammable, Flash pt. 12°C, b.p. 64-65°C
WARNING! This product can expose you to Methanol, a chemical which is known to the State of California to cause Developmental harm. For more information go to www.P65Warnings.ca.gov.