missing translation for 'onlineSavingsMsg'
Learn More

Methanol with 0.1% Formic acid, LabReady™ Solvent Blend, Honeywell Burdick & Jackson

Catalog No. 6000057
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000057 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000057 Supplier Honeywell Chemicals Supplier No. NS44520
Only null left

Chemical Identifiers

CAS 67-56-1 , 64-18-6
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 64.7°C
CAS Max % 99.9000%, 0.1000%
UN Number UN1230
Density 0.792 g/cm3
Grade Solvent Blend
Packaging Returnable Nowpak™ Ii
Quantity 20 L
Flash Point 11°C
Chemical Name or Material Methanol with 0.1% Formic acid, LabReady™ Solvent Blend
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol Solution