missing translation for 'onlineSavingsMsg'
Learn More

Methanol with 0.1% formic acid, CHROMASOLV™ LC-MS, LabReady™ Solvent Blend, Riedel-de Haen, Honeywell

$247.92 - $247.92

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60-090-293
View Documents
Honeywell Inc
346714X25L
2.5 L
Each for $247.92
Case of 4 Each for $991.68
Only null left
 
Description

Description

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CO
CH4O
MFCD00004595
methanol

Specifications

67-56-1,64-18-6
no data available
2.5 L
Glass Bottle
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
LC-MS
99.90,0.10
No data available
11°C
Liquid
MFCD00004595
CO
32.04
Videos
Safety and Handling

Safety and Handling

H225+H370

P280+P270+P264+P260+P243+P242+P241+P240+P233+P210

Danger

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'