missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, silver salt, 99%

CAS: 2386-52-9 | CH3AgO3S | 202.97 g/mol

$480.72 - $480.72

Chemical Identifiers

CAS 2386-52-9
Molecular Formula CH3AgO3S
Molecular Weight (g/mol) 202.97
MDL Number MFCD00064795
InChI Key MLKQJVFHEUORBO-UHFFFAOYSA-M
Synonym silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
PubChem CID 159658
IUPAC Name silver;methanesulfonate
SMILES CS(=O)(=O)[O-].[Ag+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC419340250
SDS View Documents
Thermo Scientific Chemicals
419340250
25 g Glass bottle
Each for $480.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2386-52-9
202.97
MLKQJVFHEUORBO-UHFFFAOYSA-M
159658
CS(=O)(=O)[O-].[Ag+]
CH3AgO3S
MFCD00064795
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
silver;methanesulfonate

Specifications

252.0°C to 256.0°C
Beige to White or Gray
25 g
CH3AgO3S
MFCD00064795
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver;methanesulfonate
159658
99%
Glass bottle
2386-52-9
Crystalline Powder
98.5% min. (Argentometry)
CH3SO3Ag
04,4
Solubility in water: soluble.
CS(=O)(=O)[O-].[Ag+]
202.97
202.97
Authentic
Methanesulfonic acid, silver salt, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a po

GHS Signal Word: Warning

EINECSNumber : 219-199-2

RTECSNumber : PB2685000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only