missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Methanesulfonic Acid

Catalog No. 471356500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
471356500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 471356500ML Supplier Sigma Aldrich Supplier No. 471356500ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methanesulfonic Acid

Specifications

CAS 75-75-2
Melting Point 17°C to 19°C
Density 1.481 g/mL at 25°C
Boiling Point 167°C (10 mmHg, lit.)
Molecular Formula CH4O3S
Refractive Index n20/D 1.429
Linear Formula CH3SO3H
MDL Number MFCD00007518
Quantity 500 mL
Molecular Weight (g/mol) 96.11 g/mol