missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonamide, 98%

Catalog No. AC296111000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296111000 100 g Glass bottle
AC296110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296111000 Supplier Thermo Scientific Chemicals Supplier No. 296111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3144-09-0 | CH5NO2S | 95.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3144-09-0
Molecular Formula CH5NO2S
Molecular Weight (g/mol) 95.12
MDL Number MFCD00007940
InChI Key HNQIVZYLYMDVSB-UHFFFAOYSA-N
Synonym methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide
PubChem CID 72879
IUPAC Name methanesulfonamide
SMILES CS(N)(=O)=O

Specifications

Melting Point 88.0°C to 92.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO2NH2
Quantity 100 g
Solubility Information Solubility in water: soluble
Formula Weight 95.12
Percent Purity 98%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material Methanesulfonamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 221-553-6

RUO – Research Use Only