missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfinic acid, sodium salt, 85%

Catalog No. AC392960100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392960100 10 g Glass bottle
AC392960500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392960100 Supplier Thermo Scientific Chemicals Supplier No. 392960100

CAS: 20277-69-4 | CH3NaO2S | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20277-69-4
Molecular Formula CH3NaO2S
Molecular Weight (g/mol) 102.09
InChI Key LYPGDCWPTHTUDO-UHFFFAOYSA-M
Synonym sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
PubChem CID 2733271
IUPAC Name sodium;methanesulfinate
SMILES CS(=O)[O-].[Na+]

Specifications

Melting Point 222.0°C to 226.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Linear Formula CH3SO2Na
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 102.09
Percent Purity 85%
Physical Form Powder
Chemical Name or Material Methanesulfinic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 243-669-6

RUO – Research Use Only