missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanesulfinic acid, sodium salt, 85%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 392960010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 20277-69-4 | |
| 102.09 | |
| sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na | |
| sodium;methanesulfinate |
| CH3NaO2S | |
| LYPGDCWPTHTUDO-UHFFFAOYSA-M | |
| 2733271 | |
| CS(=O)[O-].[Na+] |
Specifications
| Methanesulfinic acid, sodium salt | |
| Authentic | |
| Glass bottle | |
| CH3SO2Na | |
| sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na | |
| LYPGDCWPTHTUDO-UHFFFAOYSA-M | |
| sodium;methanesulfinate | |
| 2733271 | |
| 85% |
| 20277-69-4 | |
| 85% | |
| CH3NaO2S | |
| 1g | |
| Solubility in water: soluble | |
| CS(=O)[O-].[Na+] | |
| 102.09 | |
| 102.09 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 243-669-6