missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanedisulfonyl Dichloride 96.0+%, TCI America™
$411.98 - $1463.13
Chemical Identifiers
| CAS | 5799-68-8 |
|---|---|
| Molecular Formula | CH2Cl2O4S2 |
| Molecular Weight (g/mol) | 213.04 |
| MDL Number | MFCD00176854 |
| InChI Key | DKZQZHQHDMVGHE-UHFFFAOYSA-N |
| Synonym | Methylenedisulfonyl Dichloride, Methanedisulfonyl Chloride |
| PubChem CID | 79863 |
| IUPAC Name | methanedisulfonyl dichloride |
| SMILES | ClS(=O)(=O)CS(Cl)(=O)=O |
Chemical Identifiers
| 5799-68-8 | |
| 213.04 | |
| DKZQZHQHDMVGHE-UHFFFAOYSA-N | |
| 79863 | |
| ClS(=O)(=O)CS(Cl)(=O)=O |
| CH2Cl2O4S2 | |
| MFCD00176854 | |
| Methylenedisulfonyl Dichloride, Methanedisulfonyl Chloride | |
| methanedisulfonyl dichloride |
Specifications
| 5799-68-8 | |
| CH2Cl2O4S2 | |
| 5 g | |
| Methylenedisulfonyl Dichloride, Methanedisulfonyl Chloride | |
| ClS(=O)(=O)CS(Cl)(=O)=O | |
| 213.04 | |
| 213.04 | |
| Liquid |
| 138°C | |
| MFCD00176854 | |
| 3265 | |
| DKZQZHQHDMVGHE-UHFFFAOYSA-N | |
| methanedisulfonyl dichloride | |
| 79863 | |
| ≥96.0% (T) | |
| Methanedisulfonyl Dichloride |
Safety and Handling
TSCA : No