missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Methacrylic anhydride

Catalog No. 276685500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
276685500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 276685500ML Supplier Sigma Aldrich Supplier No. 276685500ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The copolymerization of methacrylic anhydride at 36.6oC produces its polymer. It can be prepared directly from the corresponding acids through the use of dehydrating agents and indirectly from the reaction of sodium salts of the acids with corresponding acid chlorides.Gelatins prepared from methayacrylamide hydrogel have high storage modulus.

Specifications

CAS 760-93-0
Density 1.035 g/mL (at 25°C)
Boiling Point 87°C (13 mmHg)
Molecular Formula C8H10O3
Refractive Index n20/D 1.453 (literature)
Linear Formula [H2C=C(CH3)CO]2 O
Quantity 500 mL
MDL Number MFCD00008586
Molecular Weight (g/mol) 154.16
Percent Purity 94%
Biological Activity Respiratory system
Show More Show Less