missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic Acid (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. M007925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M007925G 25 g
M0079500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M007925G Supplier TCI America Supplier No. M007925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-41-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002651
InChI Key CERQOIWHTDAKMF-UHFFFAOYSA-N
Synonym methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
PubChem CID 4093
ChEBI CHEBI:25219
IUPAC Name 2-methylprop-2-enoic acid
SMILES CC(=C)C(=O)O

Specifications

Color Colorless
Boiling Point 160°C
Quantity 25 g
UN Number 2531
Formula Weight 86.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methacrylic Acid (stabilized with MEHQ)
EINECSNumber (2)-1025
RTECSNumber OZ2975000
TSCA Yes