missing translation for 'onlineSavingsMsg'
Learn More

Methacrylamide 98.0+%, TCI America™

Numéro de catalogue. M007725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M007725G 25 g
M0077500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M007725G Fournisseur TCI America Code fournisseur M007725G
Il en reste null

Identifiants chimiques

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Spécifications

Melting Point 110°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 85.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methacrylamide
EINECSNumber (2)-1065
RTECSNumber UC6475000
TSCA Yes