Learn More
Methacrylamide, 98%
CAS: 79-39-0 | C4H7NO | 85.106 g/mol
Supplier: Thermo Scientific Chemicals L1501336
Description
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.
Solubility
Soluble in water (202g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
Specifications
| Methacrylamide | |
| 107°C to 112°C | |
| 215°C | |
| MFCD00008018 | |
| 605397 | |
| Soluble in water (202g/L at 20°C). | |
| CC(=C)C(=O)N | |
| 85.106 | |
| CHEBI:51759 | |
| 98% |
| 79-39-0 | |
| 1.115 | |
| C4H7NO | |
| 500 g | |
| methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide | |
| FQPSGWSUVKBHSU-UHFFFAOYSA-N | |
| 2-methylprop-2-enamide | |
| 6595 | |
| 85.11 |
Chemical Identifiers
| 79-39-0 | |
| 85.106 | |
| FQPSGWSUVKBHSU-UHFFFAOYSA-N | |
| 6595 | |
| 2-methylprop-2-enamide |
| C4H7NO | |
| MFCD00008018 | |
| methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide | |
| CHEBI:51759 | |
| CC(=C)C(=O)N |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P309+P311-P312-P330-P501c
H302-H319-H335-H371-H373
EINECSNumber : 201-202-3
RTECSNumber : UC6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only