missing translation for 'onlineSavingsMsg'
Learn More

Methacroylcholine Chloride (ca. 80% in Water) (stabilized with MEHQ), TCI America™

Catalog No. M091825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M091825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M091825G Supplier TCI America Supplier No. M091825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5039-78-1
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00060097
InChI Key RRHXZLALVWBDKH-UHFFFAOYSA-M
Synonym polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
PubChem CID 78738
IUPAC Name trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
SMILES CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 207.70
Physical Form Liquid
Chemical Name or Material Methacroylcholine Chloride (ca. 80% in Water) (stabilized with MEHQ)