missing translation for 'onlineSavingsMsg'
Learn More

Metformin Hydrochloride 98.0+%, TCI America™

Catalog No. p-7132963
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M200925G 25 g
M2009100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M200925G Supplier TCI America Supplier No. M200925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
PubChem CID 14219
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

Melting Point 221°C
Color White
Quantity 25 g
Formula Weight 165.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Metformin Hydrochloride
EINECSNumber (2)-2883
RTECSNumber DU1800000
TSCA Yes