missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metaphosphoric acid, stabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Heavy Metals (as Pb) | 50ppm max. |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Physical Form | Glassy Lump |
| Linear Formula | (HPO3)n |
| Fieser | 01,665; 16,177 |
| Merck Index | 15, 7457 |
| Solubility Information | Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol |
| Formula Weight | 79.98 |
| Percent Purity | 40 to 46% (HPO3) |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 253-433-4 |
| TSCA | TSCA |
RUO – Research Use Only