missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric acid, stabilized

Catalog No. AC219221000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219221000 100 g Glass bottle
AC219225000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC219221000 Supplier Thermo Scientific Chemicals Supplier No. 219221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max. | CAS: 37267-86-0 | HO3P | 79.979 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37267-86-0 , 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Heavy Metals (as Pb) 50ppm max.
Packaging Glass bottle
Quantity 100 g
Physical Form Glassy Lump
Linear Formula (HPO3)n
Fieser 01,665; 16,177
Merck Index 15, 7457
Solubility Information Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol
Formula Weight 79.98
Percent Purity 40 to 46% (HPO3)
Chemical Name or Material Metaphosphoric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-433-4
TSCA TSCA

RUO – Research Use Only