Learn More
Eisco™ Metamorphic Rocks Kit with Storage Box and ID Card
A fun introduction to the world of geology through a 12-piece rock kit. Each Tech Cut specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features.
Supplier:  Eisco™ ESNGKIT0007
Catalog No. S012306
Only null left
Explore available promotions
Description
Description
  • Metamorphic rocks were once another type of rock but have been transformed through high heat and pressure
  • Eisco Labs™ Tech Cut Rock and Mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Set includes twelve different metamorphic rocks: Anthracite Coal, Biotite Gneiss, Coarse Marble, Fine Marble, Phyllite, White Quartzite, Pink Quartzite, Mica Shist, Red Slate, Green Slate, Gray Slate and Soapstone
  • While each specimen will vary in shape and size, they will measure approximately 1 in. each
  • Test hardness, streak color, magnetic properties, appearance, and effervescence to identify the different specimens in this kit
  • Packaged in a box with substantial protective foam padding to store and preserve specimens
  • Ideal for home schooling or a fun home science activity, and is a great addition to any classroom or geology laboratory
  • Useful for demonstrations in identifying and classifying rocks and minerals, especially for group activities
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Metamorphic Rock
Product Suggestions
Product Suggestions