missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Metalguard™ 1880 Heavy Weight Gloves

Catalog No. 17200383
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17200383 X-Large
17200379 X-Small
17200380 Small
17200381 Medium
17200382 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17200383 Supplier Wells Lamont™ Supplier No. 1880XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High level cut-resistant Metalguard™ 1880 Heavy Weight Gloves feature ANSI Level A6 protection for use in metal handling and other applications.

  • Reinforced thumb crotch for longer life
  • Continuous knit for close secure fit
  • Ambidextrous
  • Prewashed and preshrunk
  • ANSI Level A6 cut resistance
  • Made in the USA

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F2992-15, ASTM D3884-09
Construction Seamless Knit
Coating Material Uncoated
Color Yellow, Green
Cut Resistance Rating ANSI A6
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Steel processing, metal handling, stamping, fabricating and framing
Glove Cuff Color Orange
Model 1880XL
Series Metalguard™ 1880
Yarn Gauge 7
Show More Show Less