missing translation for 'onlineSavingsMsg'
Learn More

Metaldehyde 97.0+%, TCI America™

Catalog No. M007625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M007625G 25 g
M0076500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M007625G Supplier TCI America Supplier No. M007625G

Chemical Identifiers

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00134115
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehyd
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Specifications

Melting Point 246°C
Color White
Boiling Point 65°C
Quantity 25 g
UN Number 1332
Formula Weight 176.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Metaldehyde
EINECSNumber (2)-0484
RTECSNumber XF9900000
TSCA No
Recommended Storage Refrigerator