missing translation for 'onlineSavingsMsg'
Learn More

Metaldehyde, 99%

Catalog No. AC271862500
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271862500 250 g Plastic bottle
AC271860050 5 g Glass bottle
AC271860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271862500 Supplier Thermo Scientific Chemicals Supplier No. 271862500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-62-3 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00071549
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehyd
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Specifications

Melting Point 246.0°C
Color White
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3CHO)4
Quantity 250 g
Beilstein 01, 602
Merck Index 14, 5923
Solubility Information Solubility in water: 0.02 w/w % (20°C). Other solubilities: soluble in benzene,chloroform,sparingly soluble in alcohol,ether
Sublimation Point 0112.0°C
Formula Weight 176.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Metaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Flammable solid.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
We
EINECSNumber 203-600-2
RTECSNumber AB3042000
TSCA TSCA

RUO – Research Use Only