missing translation for 'onlineSavingsMsg'
Learn More

Metaldehyde, 98%

Catalog No. AAA1529218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1529218 50 g
AAA1529230 250 g
AAA152920B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1529218 Supplier Thermo Scientific Chemicals Supplier No. A1529218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-62-3 | C8H16O4 | 176.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00071549
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehyd
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Specifications

Melting Point 192°C to 196°C (sublimation)
Flash Point 55°C (131°F)
Quantity 50 g
UN Number UN1332
Sensitivity Light sensitive
Merck Index 14,5923
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material Metaldehyde
Hazard Category H228-H301-H330
Hazard Statement GHS H Statement
H301-H330-H228
Toxic if swallowed.
Fatal if inhaled.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P284-P301+P310-P304+P340-P310-P314-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METALDEHYDE
EINECSNumber 203-600-2
RTECSNumber XF9900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only