missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Metal Draining Rack

Catalog No. S41467
Change view
Click to view available options
Positions:
32
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S41467 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S41467 Supplier Eisco™ Supplier No. CH0715B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vinyl coated metal draining rack designed to sit on the benchtop or be hung on the wall for draining/drying of labware after washing. Includes 32 integrated pegs with rubber tips. Mounting hardware not included.

  • Vinyl coated metal draining/drying rack
  • Hold up to 32 labware items at a time
  • Vinyl coated pegs with rubber tips are gentle on glassware
  • Can be used on the benchtop or hung on the wall
  • Mounting hardware not included

Warranty

1 year limited manufacturer warranty

Specifications

Autoclavable Not Autoclavable
Color White
Holds Labware
Material Vinyl-coated Metal
Positions 32
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Application) Drying Labware
For Use With (Equipment) Glassware, Plasticware
Mounting Benchtop or Wall
Quantity 1
Length (Metric) 43.8 cm
Weight (Metric) 1.18 kg
Includes Rack with 32 pegs
Product Type Draining Rack
Shelves 4 Tiers
Height (Metric) 54.6 cm
Array 8 x 4
Width (Metric) 30.5 cm
Show More Show Less