missing translation for 'onlineSavingsMsg'
Learn More

Metacresol Purple Sodium Salt, TCI America™

Catalog No. p-7135781
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00451G 1 g
S00455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00451G Supplier TCI America Supplier No. S00451G
Only null left

Chemical Identifiers

CAS 62625-31-4
Molecular Formula C21H18O5S
MDL Number MFCD00010177
Synonym m-Cresol Purple Sodium Salt, Sodium m-Cresol Purple, Sodium Metacresol Purple

Specifications

Formula Weight 404.41
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Metacresol Purple Sodium Salt