missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metacresol Purple Sodium Salt, TCI America™
Specifications
Specifications
| Formula Weight | 404.41 |
| Quantity | 1 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Metacresol Purple Sodium Salt |
Safety and Handling
| TSCA | Yes |