missing translation for 'onlineSavingsMsg'
Learn More

Meta-Phosphoric Acid, Honeywell Fluka™

Catalog No. p-7071712
Change view
Click to view available options
Quantity:
250 g
1 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000928 250 g
6000927 1 kg
6000926 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000928 Supplier Honeywell Chemicals Supplier No. 04103250G
Only null left

Lumps (glassy), 100%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Quantity 250 g
Linear Formula (HPO3)n
UN Number 3260
Chemical Name or Material Meta-Phosphoric Acid
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H314
EINECSNumber 253-433-4