missing translation for 'onlineSavingsMsg'
Learn More

Meta-Phosphoric Acid, Solstice

Catalog No. 6000927
Change view
Click to view available options
Quantity:
1 kg
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000927 1 kg
6000926 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000927 Supplier Solstice Supplier No. 041031KG

missing translation for 'validMainframeMsg'

Lumps (glassy), 100%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Quantity 1 kg
Linear Formula (HPO3)n
Chemical Name or Material Meta-Phosphoric Acid
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H314
EINECSNumber 253-433-4