Learn More
SignalChem Biotech MET (T992I), Active 50ug
Catalog No. 502574191
Encompass
Recombinant human MET (T992I) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502574191 Supplier SignalChem Biotech Supplier No. M5212DG50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human MET (T992I) (956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.