missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Met-Enkephalin

CAS: 58569-55-4573.665 g/mol

$330.11 - $992.10

Chemical Identifiers

CAS 58569-55-4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61154MB
View Documents
Thermo Scientific Chemicals
J61154MB
25 mg
Each for $330.11
Only null left
 
AAJ61154MF
View Documents
Thermo Scientific Chemicals
J61154MF
50 mg
Each for $582.55
Only null left
 
AAJ61154GPL
View Documents
Thermo Scientific Chemicals
J61154GPL
125 mg
Each for $992.10
Only null left
 
Description

Description

Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
Specifications

Chemical Identifiers

58569-55-4

Specifications

58569-55-4
Store at -20°C
573.665
25 mg
White
Powder
Met-Enkephalin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only