missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Met-Enkephalin

Catalog No. AAJ61154MB
Change view
Click to view available options
Quantity:
25 mg
50 mg
125 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61154MB 25 mg
AAJ61154MF 50 mg
AAJ61154GPL 125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61154MB Supplier Thermo Scientific Chemicals Supplier No. J61154MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58569-55-4573.665 g/mol

Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

Chemical Identifiers

CAS 58569-55-4

Specifications

Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 573.665
Product Type Met-Enkephalin
Quantity 25 mg

RUO – Research Use Only