missing translation for 'onlineSavingsMsg'
Learn More

Mesoporphyrin IX dihydrochloride, 97%

Catalog No. AA43320MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43320MA 10 mg
AA43320MF 50 mg
AA43320MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43320MA Supplier Thermo Scientific Chemicals Supplier No. 43320MA

CAS: 68938-72-7 | C34H40Cl2N4O4 | 639.62 g/mol

Mesoporphyrin IX dihydrochloride is used reactant in constructing efficient dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68938-72-7
Molecular Formula C34H40Cl2N4O4
Molecular Weight (g/mol) 639.62
MDL Number MFCD00136049
InChI Key YUIUEEROGVTICN-UHFFFAOYSA-N
Synonym mesoporphyrin ix dihydrochloride, mesoporphyrin ix, dihydrochloride, 7,12-diethyl-3,8,13,17-tetramethyl-21h,23h-porphine-2,18-dipropionic acid dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, hydrochloride 1:2, mesoporphyrin ix dihydrochloride, synthetic, 3-18-2-carboxyethyl-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl propanoic acid dihydrochloride, 3-20-2-carboxyethyl-10,15-diethyl-5,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo 16.2.1.1_,?.1?,__.1__,_? tetracosa-1,3 24 ,4,6,8,10,12,14,16 22 ,17,19-undecaen-4-yl propanoic acid dihydrochloride
PubChem CID 111858
SMILES [H+].[H+].[Cl-].[Cl-].CCC1=C(C)C2=CC3=C(CC)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C

Specifications

Quantity 10 mg
Solubility Information Soluble in water.
IUPAC Name 3-[18-(2-carboxyethyl)-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl]propanoic acid;dihydrochloride
Formula Weight 639.62
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Mesoporphyrin IX dihydrochloride
EINECSNumber 273-141-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only