missing translation for 'onlineSavingsMsg'
Learn More

meso-Tetraphenylporphine, low chlorine

Catalog No. AAA1826103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1826103 1 g
AAA1826106 5 g
AAA1826114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1826103 Supplier Thermo Scientific Chemicals Supplier No. A1826103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 917-23-7 | C44H30N4 | 614.75 g/mol

meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 917-23-7
Molecular Formula C44H30N4
Molecular Weight (g/mol) 614.75
MDL Number MFCD00011680
InChI Key AQPPOLXYUQPDOD-UHFFFAOYSA-N
Synonym tetraphenylporphyrin, 5,10,15,20-tetraphenylporphyrin, meso-tetraphenylporphine, tetraphenylporphine, 5,10,15,20-tetraphenylporphine, meso-tetraphenylporphyrin, 21h,23h-porphine, 5,10,15,20-tetraphenyl, 5,10,15,20-tetraphenyl-21h,23h-porphine, meso-tetraphenylporphyrine, tetraphenyl porphine
PubChem CID 70186
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 449°C to 451°C
Quantity 1 g
Beilstein 379542
Solubility Information Insoluble in water.
IUPAC Name 5,10,15,20-tetraphenyl-21,22-dihydroporphyrin
Formula Weight 614.71
Chemical Name or Material meso-Tetraphenylporphine, low chlorine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-025-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only