missing translation for 'onlineSavingsMsg'
Learn More

meso-Hydrobenzoin 98.0+%, TCI America™

Catalog No. p-7130741
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
H08165G 5 g
H081625G 25 g
2 options

Catalog No. H08165G

Supplier: TCI America H08165G

Only null left

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name 1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material meso-Hydrobenzoin