missing translation for 'onlineSavingsMsg'
Learn More

meso-Butane-1,2,3,4-tetracarboxylic Dianhydride 96.0+%, TCI America™

Catalog No. B17705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17705G 5 g
B177025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17705G Supplier TCI America Supplier No. B17705G

Chemical Identifiers

CAS 17309-39-6
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00671500
InChI Key OLQWMCSSZKNOLQ-WUCPZUCCSA-N
PubChem CID 54156817
IUPAC Name 3-[(3S)-2,5-dioxooxolan-3-yl]oxolane-2,5-dione
SMILES C1C(C(=O)OC1=O)C2CC(=O)OC2=O

Specifications

Color White
Quantity 5 g
Formula Weight 198.13
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material meso-Butane-1,2,3,4-tetracarboxylic Dianhydride