missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Diphenylethylenediamine 97.0+%, TCI America™

Catalog No. D39301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39301G 1 g
D39305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39301G Supplier TCI America Supplier No. D39301G
Only null left

Chemical Identifiers

CAS 951-87-1
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00274328
InChI Key PONXTPCRRASWKW-OKILXGFUSA-N
PubChem CID 6931234
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.30
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material meso-1,2-Diphenylethylenediamine