missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Diphenyl-1,2-ethanediol, 95%

Catalog No. AC247680050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC247680050 5 g Glass bottle
AC247680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC247680050 Supplier Thermo Scientific Chemicals Supplier No. 247680050

CAS: 579-43-1 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136.0°C to 137.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(OH)CH(OH)C6H5
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 214.26
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material meso-1, 2-Diphenyl-1, 2-ethanediol
Show More Show Less

RUO – Research Use Only