missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Oxide, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023055
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023055 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023055 Supplier Merck Emd Millipore Supplier No. RTC00009720ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 141-79-7
Melting Point -41.5°C (literature)
Density 0.858 g/mL (at 25°C (literature))
Boiling Point 130°C (literature)
Flash Point 31°C (Not Applicable)
Molecular Formula C6H10O
Refractive Index n20/D 1.442 (literature)
Linear Formula (CH3)2 C=CHCOCH3
MDL Number MFCD00008900
Quantity 20 mL
Synonym Mesityl oxide; 4-Methyl-3-penten-2-one
Formula Weight 98.14
Grade Certified Reference Material
Show More Show Less