missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Oxide, Approx. 98%, Spectrum™ Chemical

Catalog No. 18604334
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604334 100 mL Amber Glass Bottle
18604335 500 mL Amber Glass Bottle
18604336 2.5 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604334 Supplier Spectrum Chemical Mfg Cor Supplier No. M2491100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

M2491, 141-79-7, C6H10O

Spectrum™ Chemical Mesityl Oxide is a colorless compound with a strong peppermint scent. It is known to be a volatile liquid. Mesityl oxide is produced under the same conditions of isophorone. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(C)=CC(C)=O

Specifications

CAS Max % 100%
Boiling Point 130°C (266°F)
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN1229
Percent Purity ∼98%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Mesityl Oxide