missing translation for 'onlineSavingsMsg'
Learn More

Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one

Catalog No. AAA15111AU
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15111AU 1000 mL
AAA15111AE 100 mL
AAA15111AK 250 mL
AAA15111AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA15111AU Supplier Thermo Scientific Chemicals Supplier No. A15111AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 141-79-7 | C6H10O | 98.145 g/mol

Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008900
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
Synonym mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
PubChem CID 8858
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(=CC(=O)C)C

Specifications

Melting Point -59°C
Density 0.856
Boiling Point 129°C to 130°C
Flash Point 30°C (86°F)
Odor honigähnlich
Assay Percent Range ≥90%
Refractive Index 1.444
Quantity 1000 mL
UN Number UN1229
Beilstein 1361550
Merck Index 14,5908
Solubility Information Soluble in water(30g/L).
Formula Weight 98.15
Percent Purity ≥90%
Physical Form Liquid
Assay remainder 4-methyl-4-penten-2-one
Chemical Name or Material Mesityl oxide
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MESITYL OXIDE
EINECSNumber 205-502-5
RTECSNumber SB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only