missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Chloride 97.0+%, TCI America™

Supplier:  TCI America M02745G

Catalog No. M02745G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Mesityl Chloride
Colorless
5 g
C9H11Cl
2-Chloromesitylene, 2-Chloro-1,3,5-trimethylbenzene
CC1=CC(C)=C(Cl)C(C)=C1
154.64
154.64
1667-04-5
206°C
Clear Liquid at 20°C
MFCD00045173
WDZACGWEPQLKOM-UHFFFAOYSA-N
2-chloro-1,3,5-trimethylbenzene
15462
≥97.0% (GC)

Chemical Identifiers

1667-04-5
154.64
WDZACGWEPQLKOM-UHFFFAOYSA-N
15462
CC1=CC(C)=C(Cl)C(C)=C1
C9H11Cl
MFCD00045173
2-Chloromesitylene, 2-Chloro-1,3,5-trimethylbenzene
2-chloro-1,3,5-trimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions