Learn More
Labconco™ Mesh Flask Sleeve
Mesh sleeves act as a buffer, absorbing impacts and bumps that could crack or shatter flasks during handling and use.
Supplier:  Labconco™ 7121723
 View more versions of this product
Catalog No. 16-985-657
Only null left
Explore available promotions
Description
Description
  • The mesh texture can improve grip, making it less likely to slip out of your hands.
  • This is particularly useful when wearing gloves or working in cold environments.
Specifications
Specifications
15. 88 cm
Flask Sleeve
Product Suggestions
Product Suggestions