Learn More
Labconco™ Mesh Flask Sleeve
Catalog No. 16985662
Change view
Click to view available options
Height (Metric):
4.76 cm
5.4 cm
6.03 cm
9.21 cm
10.48 cm
12.07 cm
13.3 cm
15. 88 cm
20.96 cm
34.9 cm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
16985662 4.76 cm
16985660 12.07 cm
16985655 34.9 cm
16985663 9.21 cm
16985664 5.4 cm
16985661 6.03 cm
16985659 10.48 cm
16985658 13.3 cm
16985656 20.96 cm
16985657 15. 88 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 16985662 Supplier Labconco™ Supplier No. 7121711
Mesh sleeves act as a buffer, absorbing impacts and bumps that could crack or shatter flasks during handling and use.
  • The mesh texture can improve grip, making it less likely to slip out of your hands.
  • This is particularly useful when wearing gloves or working in cold environments.
Specifications
Height (Metric) 4.76 cm
Product Type Flask Sleeve