Learn More
Mesa Labs Prospore™ Self-Contained Biological Indicator
Catalog No. 120011
Encompass
Change view
Click to view available options
Quantity:
Pack of 10
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120011 Pack of 10
120012 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 120011 Supplier Mesa Labs Supplier No. PS510
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for validating steam sterilization of liquids
Safe, easy-to-use, self-contained biological indicator used to validate or monitor steam sterilization in hospitals and pharmaceutical facilities.
  • No activation is required—simply expose ampule sterilizer and incubate afterwards
  • Sealed Prospore ampules contain Geobacillus stearothermophilus spores suspended in growth media
  • Bromocresol purple pH indicator added to assist in detecting spore growth
  • Outgrowth of spores decreases pH, causing a color change from purple to yellow
  • Standard population is 105 per unit
  • Results available in 48 hours
  • 4mL ampules
Order Info
For 121°C steam sterilization cycles only. A control test should be used to verify spore viability.
Specifications
For Use With (Application) Validating steam sterilization of liquids
Temperature (English) 35.6°F to 46.4°F
Temperature (Metric) 2°C to 8°C
Population 105
Quantity Pack of 10