missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MES, Sodium Salt, Purified, J.T. Baker™
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Formula Weight | 217.2 |
| Grade | Purified |
| Chemical Name or Material | MES, Sodium Salt |