missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma MES SDS Running Buffer Powder

Catalog No. 11580599
Change view
Click to view available options
Physical Form:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Physical Form
11580599 Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11580599 Supplier MilliporeSigma Supplier No. MPMES

MES SDS running buffer powder for mPAGE™ Bis-Tris gels

  • MES SDS running buffer powder for mPAGEBis-Tris gels.
  • 5 packets.
  • Each paclet makes 1 liter of running buffer.
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Specifications

Chemical Name or Material MES SDS running buffer powder for mPAGE™ Bis-Tris gels
Physical Form Powder
Quantity 5 Pk.