missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES hydrate, 99+%

CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol

$282.45 - $448.38

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5647230
View Documents
Thermo Scientific Chemicals
H5647230
250 g
Each for $282.45
Only null left
 
AAH5647236
View Documents
Thermo Scientific Chemicals
H5647236
500 g
Each for $448.38
Only null left
 
Description

Description

MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water. Slightly soluble in organic solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1266615-59-1
195.23
SXGZJKUKBWWHRA-UHFFFAOYSA-N
2-(morpholin-4-yl)ethane-1-sulfonic acid
C6H13NO4S
MFCD00149409
2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
OS(=O)(=O)CCN1CCOCC1

Specifications

1266615-59-1
C6H13NO4S
250 g
14,5902
Soluble in water. Slightly soluble in organic solvents.
OS(=O)(=O)CCN1CCOCC1
195.23
≥99%
∽308°C (decomposition)
MFCD00149409
141319
2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
SXGZJKUKBWWHRA-UHFFFAOYSA-N
2-(morpholin-4-yl)ethane-1-sulfonic acid
195.24 (Anhydrous)
MES hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 224-632-3

RTECSNumber : QE3479500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only