missing translation for 'onlineSavingsMsg'
Learn More

Meropenem Trihydrate 98.0+%, TCI America™

Supplier:  TCI America M22795G

 View more versions of this product

Catalog No. M22795G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Meropenem Trihydrate
Yellow
MFCD08600005
VRWIQGNHNITILX-OBZXMJSBSA-N
(4R,5S,6S)-3-[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;trihydrochloride
129893494
≥98.0% (HPLC,T)
119478-56-7
C17H28Cl3N3O5S
5 g
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=O)N(C)C)C(=O)O)C(C)O.Cl.Cl.Cl
492.837
383.46
Crystalline Powder

Chemical Identifiers

119478-56-7
492.837
VRWIQGNHNITILX-OBZXMJSBSA-N
(4R,5S,6S)-3-[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;trihydrochloride
C17H28Cl3N3O5S
MFCD08600005
129893494
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=O)N(C)C)C(=O)O)C(C)O.Cl.Cl.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CL5446507

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions