missing translation for 'onlineSavingsMsg'
Learn More

Meropenem Trihydrate 98.0+%, TCI America™

Catalog No. M22791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22791G 1 g
M22795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22791G Supplier TCI America Supplier No. M22791G
Only null left

Chemical Identifiers

CAS 119478-56-7
Molecular Formula C17H28Cl3N3O5S
Molecular Weight (g/mol) 492.837
MDL Number MFCD08600005
InChI Key VRWIQGNHNITILX-OBZXMJSBSA-N
PubChem CID 129893494
IUPAC Name (4R,5S,6S)-3-[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;trihydrochloride
SMILES CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=O)N(C)C)C(=O)O)C(C)O.Cl.Cl.Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 383.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Meropenem Trihydrate
RTECSNumber CL5446507
TSCA No
Recommended Storage Refrigerator