missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meropenem trihydrate, 98-101%
Description
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
Solubility
Soluble in aqueous solution at approximately 5mg/ml
Notes
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
Solubility
Soluble in aqueous solution at approximately 5mg/ml
Notes
Specifications
Specifications
| Type | Meropenem Trihydrate |
| Physical Form | Powder |
| Color | White to Yellow |
| Quantity | 50 mg |
| Flash Point | >110°C (230°F) |
| Boiling Point | 627°C |
| Merck Index | 14,5900 |
| Solubility Information | Soluble in aqueous solution at approximately 5mg/mL |
| Formula Weight | 383.46 |
| Percent Purity | 98 to 101% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| RTECSNumber | CL5446509 |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only